Order Entry
Czech Republic
ContactUsLinkComponent
Ciprofloxacin hydrochloride monohydrate ≥98% (by HPLC)
Ciprofloxacin hydrochloride monohydrate ≥98% (by HPLC)
Catalog # ENZOALX380288G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ciprofloxacin hydrochloride monohydrate ≥98% (by HPLC)
Catalog # ENZOALX380288G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :385.82 g/mol
Storage Temperature:Freezer
CAS Number:86393-32-0
Formula:C₁₇H₁₈FN₃O₃·HCl·H₂O
MDL Number:MFCD00242856