Order Entry
Czech Republic
ContactUsLinkComponent
Leptomycin B 0.2 mg/ml ≥98% (by HPLC)
Leptomycin B 0.2 mg/ml ≥98% (by HPLC)
Catalog # ENZOALX380100C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Leptomycin B 0.2 mg/ml ≥98% (by HPLC)
Catalog # ENZOALX380100C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Seq:
    1286

Specifications

About this item

 :
 :540.73 g/mol
Storage Temperature:Freezer
CAS Number:87081-35-4
Formula:C₃₃H₄₈O₆
MDL Number:MFCD06795848