Antimycin A (from Streptomyces species) ≥90% (by HPLC)
:1397-94-0
:
:C₂₈H₄₀N₂O₉
:MFCD01779723
:548.63 g/mol
:Freezer
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Molykote® High Vaccum Grease
: DOW CORNING