Order Entry
Czech Republic
ContactUsLinkComponent
 
Hygromycin B (from Streptomyces hygroscopicus) ≥80% (by HPLC), liquid
  ENZOALX380059UM01
 :  
Hygromycin B (from Streptomyces hygroscopicus) ≥80% (by HPLC), liquid
  ENZOALX380059UM01
 :  

 

  • Pk:
    1 Mu
  • Seq:
    6200

 

 

 :
 :527.5 g/mol
 :Refrigerator
 :31282-04-9
 :C₂₀H₃₇N₃O₁₃
 :MFCD06795479