Order Entry
Czech Republic
ContactUsLinkComponent
Kanamycin sulphate
Kanamycin sulphate
Catalog # ENZOALX380049G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Kanamycin sulphate
Catalog # ENZOALX380049G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :582.6 g/mol
EINECS:246-933-9
 :GHS08
Storage Temperature:Ambient
CAS Number:25389-94-0
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253