Order Entry
Czech Republic
ContactUsLinkComponent
Nourseothricin sulphate (from Streptomyces noursei) ≥85%
Nourseothricin sulphate (from Streptomyces noursei) ≥85%
Catalog # ENZOALX380025M005
Supplier:  Enzo Life Sciences
CAS Number:  
Nourseothricin sulphate (from Streptomyces noursei) ≥85%
Catalog # ENZOALX380025M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1359.47 g/mol
Storage Temperature:Refrigerator
CAS Number:96736-11-7
 :MFCD06795125
Formula:C₃₁H₅₈N₁₂O₁₀·C₁₉H₃₄N₈O₈·H₂SO₄
MDL Number:MFCD00165013