Order Entry
Czech Republic
ContactUsLinkComponent
 
 
3,3'-Methylenediindole ≥98% (by HPLC)
  ENZOALX350370M500
 :  
undefined
 
3,3'-Methylenediindole ≥98% (by HPLC)
  ENZOALX350370M500
 :  

 

  • Pk:
    500 mg
  • Seq:
    2725

 

 

  • Purity:
    ≥98% (HPLC)

 

 

 :
 :246.31 g/mol
 :GHS07
 :Freezer
 :1968-05-4
 :42494-73-5
 :C₁₇H₁₄N₂
 :MFCD00195766