Auraptene ≥95% (by UPLC)
Catalog # ENZOALX350361M025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Purity:≥95% (UPLC)
- Identity:Determined by NMR
- Source:Isolated from citrus fruit
Specification Test Results
About this item
:
:298.4 g/mol
Storage Temperature:Freezer
CAS Number:495-02-3
Formula:C₁₉H₂₂O₃
MDL Number:MFCD00075948