Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Roquefortine C
Roquefortine C ≥95% (by HPLC)
Roquefortine C ≥95% (by HPLC)
Roquefortine C ≥95% (by HPLC)
Catalog # ENZOALX350342MC05
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Roquefortine C ≥95% (by HPLC)
Catalog # ENZOALX350342MC05
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
0,5 mg
Seq:
2035
Specifications
About this item
Product Details
:
:
389.5 g/mol
CAS Number:
58735-64-1
Formula:
C₂₂H₂₃N₅O₂
Product Details