
Harpagoside ≥98% (by HPLC)
Catalog # ENZOALX350333M025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Purity:≥98% by HPLC
- Source:Isolated from Scrophularia ningpoensis Hemsl (Figwort)
Specification Test Results
About this item
:
:494.5 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:19210-12-9
Formula:C₂₄H₃₀O₁₁
MDL Number:MFCD00017415