Order Entry
Czech Republic
ContactUsLinkComponent
(-)-Arctigenin (from Arctium lappa) ≥97% (by HPLC)
(-)-Arctigenin (from Arctium lappa) ≥97% (by HPLC)
Catalog # ENZOALX350312M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
(-)-Arctigenin (from Arctium lappa) ≥97% (by HPLC)
Catalog # ENZOALX350312M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :372.4 g/mol
Storage Temperature:Freezer
CAS Number:7770-78-7
Formula:C₂₁H₂₄O₆
MDL Number:MFCD00870597