Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-Roscovitine
(S)-Roscovitine ≥98% (by HPLC)
(S)-Roscovitine ≥98% (by HPLC)
(S)-Roscovitine ≥98% (by HPLC)
Catalog # ENZOALX350293M001
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
(S)-Roscovitine ≥98% (by HPLC)
Catalog # ENZOALX350293M001
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
354.5 g/mol
CAS Number:
186692-45-5
Formula:
C₁₉H₂₆N₆O
Product Details