Resibufogenin (from Bufonis venom) ≥98% (by HPLC)
Catalog # ENZOALX350286M010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:384.5 g/mol
Storage Temperature:Refrigerator
CAS Number:465-39-4
Formula:C₂₄H₃₂O₄
MDL Number:MFCD00210361