Order Entry
Czech Republic
ContactUsLinkComponent
(S)-(+)-Abscisic acid cis- and trans-mixture (from Curvularia lunata) ≥97% (by HPLC)
(S)-(+)-Abscisic acid cis- and trans-mixture (from Curvularia lunata) ≥97% (by HPLC)
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
(S)-(+)-Abscisic acid cis- and trans-mixture (from Curvularia lunata) ≥97% (by HPLC)
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Purity:
    ≥97% by HPLC
  • Source:
    Isolated from Curvularia lunata

Specification Test Results

Frequently Bought Together