Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Butein
Butein ≥98% (by HPLC)
Butein ≥98% (by HPLC)
Butein ≥98% (by HPLC)
Catalog # ENZOALX350246M010
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Butein ≥98% (by HPLC)
Catalog # ENZOALX350246M010
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
272.26 g/mol
CAS Number:
487-52-5
Formula:
C₁₅H₁₂O₅
MDL Number:
MFCD00017300
Product Details