ContactUsLinkComponent
Chlortetracycline hydrochloride
  ENZOALX350238G005
 :  
Chlortetracycline hydrochloride
  ENZOALX350238G005
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
 :
 :515.34 g/mol
 :200-591-7
 :GHS06
 :GHS08
 :Refrigerator
 :64-72-2
 :C₂₂H₂₃ClN₂O₈·HCl
 :MFCD26142699