Gramicidin A (from Bacillus brevis) ≥90% (by HPLC)
:11029-61-1
:11029-61-01
:C₉₉H₁₄₀N₂₀O₁₇
:MFCD00466944
:1882.3 g/mol
:Freezer
Frequently Bought Together
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Sampling scoops, LaboPlast®
: BURKLE
VWR® Media Bottles, Square, PETG
: VWR Collection
Cryogenic tubes, CryoTubes™, Nunc™
: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
Immunology plates, 384-well, MaxiSorp™, Nunc™
: Thermo Fisher Scientific
Bottle hdpe 250ml 18mg sod thio 0.125ml 1 * 25 items
: LAB3 OWN PRODUCTION
Cap sealing mats, Matrix SepraSeal® and DuraSeal®
: THERMO MATRIX TECHNOLOGIES
VWR®, Cryo Boxes
: VWR Collection
Blotting transfer papers
: Ahlstrom-Munksjö