Order Entry
Czech Republic
ContactUsLinkComponent
Ferutinin ≥98% (by HPLC), white to off-white solid, high purity
Ferutinin ≥98% (by HPLC), white to off-white solid, high purity
Catalog # ENZOALX350098M005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Ferutinin ≥98% (by HPLC), white to off-white solid, high purity
Catalog # ENZOALX350098M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98% by HPLC
  • Source:
    Semisynthetic

Specification Test Results

About this item

 :
 :358.5 g/mol
Storage Temperature:Refrigerator
CAS Number:41743-44-6
Formula:C₂₂H₃₀O₄