Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Fenofibrate ≥98% (by HPLC)
Catalog # ENZOALX270481G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Irritant
Fenofibrate ≥98% (by HPLC)
Catalog # ENZOALX270481G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :360.83 g/mol
Merck Index:14,03978
EINECS:256-376-3
 :GHS07
Storage Temperature:Freezer
CAS Number:49562-28-9
Formula:C₂₀H₂₁ClO₄
MDL Number:MFCD00133314