Order Entry
Czech Republic
ContactUsLinkComponent
Ciprofibrate ≥98% (by HPLC)
Health
Ciprofibrate ≥98% (by HPLC)
Catalog # ENZOALX270475M025
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Health
Ciprofibrate ≥98% (by HPLC)
Catalog # ENZOALX270475M025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :289.15 g/mol
Merck Index:14,02313
EINECS:257-744-6
 :GHS08
Storage Temperature:Freezer
CAS Number:52214-84-3
Formula:C₁₃H₁₄Cl₂O₃
MDL Number:MFCD00467135