Order Entry
Czech Republic
ContactUsLinkComponent
AS-604850 ≥98% (by HPLC and UV method)
AS-604850 ≥98% (by HPLC and UV method)
Catalog # ENZOALX270461M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
AS-604850 ≥98% (by HPLC and UV method)
Catalog # ENZOALX270461M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :285.2 g/mol
CAS Number:648449-76-7
Formula:C₁₁H₅F₂NO₄S
MDL Number:MFCD19705528