Order Entry
Czech Republic
ContactUsLinkComponent
PQ401 ≥98% (by HPLC and UV method)
PQ401 ≥98% (by HPLC and UV method)
Catalog # ENZOALX270459M005
Supplier:  Enzo Life Sciences
CAS Number:  
PQ401 ≥98% (by HPLC and UV method)
Catalog # ENZOALX270459M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :341.8 g/mol
Storage Temperature:Freezer
CAS Number:196868-63-0
Formula:C₁₈H₁₆ClN₃O₂
MDL Number:MFCD00160558