Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sphingolactone-24
Sphingolactone-24 ≥95%
Sphingolactone-24 ≥95%
Sphingolactone-24 ≥95%
Catalog # ENZOALX270453M001
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Sphingolactone-24 ≥95%
Catalog # ENZOALX270453M001
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
323.4 g/mol
Storage Temperature:
Freezer
CAS Number:
881177-99-7
Formula:
C₁₈H₂₉NO₄
Product Details