Order Entry
Czech Republic
ContactUsLinkComponent
AMI-1 ≥98% (by 1H-NMR, HPLC)
AMI-1 ≥98% (by 1H-NMR, HPLC)
Catalog # ENZOALX270440M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
AMI-1 ≥98% (by 1H-NMR, HPLC)
Catalog # ENZOALX270440M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :548.45 g/mol
CAS Number:20324-87-2 | 134-47-4 (free base)
Formula:C₂₁H₁₄N₂Na₂O₉S₂
MDL Number:MFCD00068249