Order Entry
Czech Republic
ContactUsLinkComponent
1,1-Dimethylbiguanide hydrochloride ≥97%
1,1-Dimethylbiguanide hydrochloride ≥97%
Catalog # ENZOALX270432G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
1,1-Dimethylbiguanide hydrochloride ≥97%
Catalog # ENZOALX270432G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :165.625 g/mol
CAS Number:1115-70-4
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582