Order Entry
Czech Republic
ContactUsLinkComponent
Rockout ≥98% (by HPLC)
Rockout ≥98% (by HPLC)
Catalog # ENZOALX270429M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Rockout ≥98% (by HPLC)
Catalog # ENZOALX270429M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :194.2 g/mol
Storage Temperature:Freezer
CAS Number:7272-84-6
Formula:C₁₃H₁₀N₂