Order Entry
Czech Republic
ContactUsLinkComponent
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Catalog # ENZOALX270425M001
Supplier:  Enzo Life Sciences
CAS Number:  
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Catalog # ENZOALX270425M001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :351.4 g/mol
Formula:C₁₉H₁₇N₃O₄