Order Entry
Czech Republic
ContactUsLinkComponent
EB-47 dihydrochloride dihydrate
EB-47 dihydrochloride dihydrate
Catalog # ENZOALX270383M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
EB-47 dihydrochloride dihydrate
Catalog # ENZOALX270383M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :646.5 g/mol
Formula:C₂₄H₂₇N₉O₆·2HCl·2H₂O