Order Entry
Czech Republic
ContactUsLinkComponent
Tryptanthrin ≥98% (by 1H-NMR, HPLC)
Tryptanthrin ≥98% (by 1H-NMR, HPLC)
Catalog # ENZOALX270360M005
Supplier:  Enzo Life Sciences
CAS Number:  
Tryptanthrin ≥98% (by 1H-NMR, HPLC)
Catalog # ENZOALX270360M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :248.2362 g/mol
CAS Number:13220-57-0
Formula:C₁₅H₈N₂O₂
MDL Number:MFCD00012073