Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ubiquinone-10
Ubiquinone-10 ≥90% (by HPLC)
Ubiquinone-10 ≥90% (by HPLC)
Ubiquinone-10 ≥90% (by HPLC)
ENZOALX270295M002
:
ENZO LIFE SCIENCES
:
Ubiquinone-10 ≥90% (by HPLC)
ENZOALX270295M002
:
ENZO LIFE SCIENCES
:
Pk:
2 mg
Seq:
2145
:
:
606-06-4