1-Methyl-3,4-bis(3-indolyl)maleimide ≥98%
Catalog # ENZOALX270053M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:341.37 g/mol
CAS Number:113963-68-1
Formula:C₂₁H₁₅N₃O₂
MDL Number:MFCD00236433