
L-α-Phosphatidylinositol-3,4,5-P3 ·7Na ≥98% (by TLC)
Catalog # ENZOBMLPH1070100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Seq:1286
Specifications
Specification Test Results
- Purity:≥98% (TLC)
- Formulation:Lyophilised
Specification Test Results
About this item
:
:1204.8 g/mol
:GHS07
Storage Temperature:Freezer
:383907-36-6
Formula:C₄₁H₇₅O₂₂P₄·7Na