Order Entry
Czech Republic
ContactUsLinkComponent
Leukotriene E4-20,20,20-d3 ≥98% (by HPLC)
Flammable
Toxic
Health
Leukotriene E4-20,20,20-d3 ≥98% (by HPLC)
Catalog # ENZOBMLLE5040050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Flammable
Toxic
Health
Leukotriene E4-20,20,20-d3 ≥98% (by HPLC)
Catalog # ENZOBMLLE5040050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Formulation:
    Solution in methanol/ammonium acetate buffer, 70:30, pH 5.6
  • Concentration:
    50 µg/ml
  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :442.6 g/mol
UN:1992
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
 :75715-89-8
Formula:C₂₃H₃₄D₃NO₅S