Order Entry
Czech Republic
ContactUsLinkComponent
N-Arachidonoyl-L-phenylalanine ≥98% (by TLC)
N-Arachidonoyl-L-phenylalanine ≥98% (by TLC)
Catalog # ENZOBMLFA0400025
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
N-Arachidonoyl-L-phenylalanine ≥98% (by TLC)
Catalog # ENZOBMLFA0400025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :451.6 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
Formula:C₂₉H₄₁NO₃