Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥95% (by HPLC), CELLESTIAL®, Ultrapure fluorescent dye
Catalog # ENZOENZ52404
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥95% (by HPLC), CELLESTIAL®, Ultrapure fluorescent dye
Catalog # ENZOENZ52404
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :350.25 g/mol
EINECS:249-186-7
 :GHS07
Storage Temperature:Freezer
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681