Order Entry
Czech Republic
ContactUsLinkComponent
DiBAC₄(3) ≥98% (by HPLC), Ultrapure membrane stain, for histology
DiBAC₄(3) ≥98% (by HPLC), Ultrapure membrane stain, for histology
Catalog # ENZOENZ52205
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
DiBAC₄(3) ≥98% (by HPLC), Ultrapure membrane stain, for histology
Catalog # ENZOENZ52205
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :516.63 g/mol
CAS Number:70363-83-6
Formula:C₂₇H₄₀N₄O₆
MDL Number:MFCD07437857