Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhod-2 AM (Bromide)
Rhod-2 AM (Bromide) ≥90% (by HPLC), Ultrapure
Rhod-2 AM (Bromide) ≥90% (by HPLC), Ultrapure
Rhod-2 AM (Bromide) ≥90% (by HPLC), Ultrapure
ENZOENZ52010
:
ENZO LIFE SCIENCES
:
Rhod-2 AM (Bromide) ≥90% (by HPLC), Ultrapure
ENZOENZ52010
:
ENZO LIFE SCIENCES
:
Pk:
1 mg
Seq:
2070
:
:
1124.0 g/mol
:
Freezer
:
145037-81-6
:
C₅₂H₅₉BrN₄O₁₉