Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TN-16
TN-16 ≥97% (by HPLC)
TN-16 ≥97% (by HPLC)
TN-16 ≥97% (by HPLC)
Catalog # ENZOBMLT1200050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
TN-16 ≥97% (by HPLC)
Catalog # ENZOBMLT1200050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
CAS Number:
33016-12-5
Product Details