Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
BML-260
BML-260 ≥98% (by TLC)
BML-260 ≥98% (by TLC)
Catalog # ENZOBMLPR1190010
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
BML-260 ≥98% (by TLC)
Catalog # ENZOBMLPR1190010
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
341.4 g/mol
Storage Temperature:
Ambient
CAS Number:
101439-76-3
Formula:
C₁₇H₁₁NO₃S₂
Product Details