PF-356231 ≥98% (by TLC)
Catalog # ENZOBMLPI1550005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:428.5 g/mol
Storage Temperature:Freezer
CAS Number:766536-21-4
Formula:C₂₅H₂₀N₂O₃S