BML-244 ≥98% (by TLC)
:104062-70-6
:
:C₁₁H₂₁NO₃
:215.3 g/mol
:Freezer
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Serological pipettes, Stripette™
: Corning
VWR®, Petri Dish, PS
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Transfer Pipettes, High Performance
: VWR Collection