Order Entry
Czech Republic
ContactUsLinkComponent
8-Methoxymethyl-IBMX ≥98% (by TLC)
8-Methoxymethyl-IBMX ≥98% (by TLC)
Catalog # ENZOBMLPD1500010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
8-Methoxymethyl-IBMX ≥98% (by TLC)
Catalog # ENZOBMLPD1500010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :266.3 g/mol
Storage Temperature:Ambient
CAS Number:78033-08-6
Formula:C₁₂H₁₈N₄O₃