EHNA ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:51350-19-7
:
Formula:C₁₄H₂₃N₅O·HCl
:313.8 g/mol
Storage Temperature:Ambient
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Self-standing centrifuge tubes, 50 ml, Corning®
Supplier: Corning
Round bottom high clarity polypropylene tube, disposable, Falcon®
Supplier: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, Low-Binding
Supplier: VWR Collection
96-Well cell culture plates, Falcon®
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
UV microplates, 96-well, Corning®
Supplier: Corning
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection