WEB-2086 ≥98% (by HPLC)
Catalog # ENZOBMLL1370005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:456 g/mol
Storage Temperature:Refrigerator
CAS Number:105219-56-5
Formula:C₂₂H₂₂ClN₅O₂S