Order Entry
Czech Republic
ContactUsLinkComponent
12(S)-Hydroperoxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HPETE) ≥98%
12(S)-Hydroperoxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HPETE) ≥98%
Catalog # ENZOBMLHP0120050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
12(S)-Hydroperoxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HPETE) ≥98%
Catalog # ENZOBMLHP0120050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

About this item

 :
 :336.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
CAS Number:71774-10-2
Formula:C₂₀H₃₂O₄