BML-266 ≥95% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:96969-83-4
:
Formula:C₃₀H₂₆N₂O₄
:478.5 g/mol
Storage Temperature:Freezer
Frequently Bought Together
Ice pans, Bel-Art Magic Touch 2™
Supplier: Bel-Art Products, a Part of SP
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Supplier: Thermo Fisher Scientific
Racks and lids for storage 2D barcoded tubes, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Polypropylene microplates, 96- and 384-well
Supplier: EPPENDORF
Petri dishes with absorbent pads
Supplier: Simport Scientific
Cryo vial closure colour coders, Nalgene®
Supplier: Thermo Fisher Scientific
Stomacher Model 400 Eco Bag - TVC. Dimensions 155 x 200 mm 1 * 500 items
Supplier: Seward (FermionX)
Cellulose chromatography papers, Whatman™
Supplier: Whatman products (Cytiva)
Sample cups, SeraNest™
Supplier: Simport Scientific
Cryoboxes, B47/B67/B77/B97-series
Supplier: GLW
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Deepwell Plates 96-well, 500 µl
Supplier: EPPENDORF
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling