Order Entry
Czech Republic
ContactUsLinkComponent
Lovastatin hydroxy acid sodium ≥97% (by HPLC)
Lovastatin hydroxy acid sodium ≥97% (by HPLC)
Catalog # ENZOBMLG2450005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Lovastatin hydroxy acid sodium ≥97% (by HPLC)
Catalog # ENZOBMLG2450005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :444.54 g/mol
Storage Temperature:Freezer
CAS Number:75225-50-2
Formula:C₂₄H₃₇O₆·Na
MDL Number:MFCD00870475