Order Entry
Czech Republic
ContactUsLinkComponent
Mevastatin sodium ≥98% (by HPLC)
Mevastatin sodium ≥98% (by HPLC)
Katalogové číslo ENZOBMLG2380005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Mevastatin sodium ≥98% (by HPLC)
Katalogové číslo ENZOBMLG2380005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :430.51 g/mol
Melting Pt:44…45 °C
Storage Temperature:Freezer
CAS Number:99782-89-5
Formula:C₂₃H₃₅O₆·Na