Order Entry
Czech Republic
ContactUsLinkComponent
 
K-252b ≥98% (by HPLC)
  ENZOBMLEI2340100
 :  
undefined
K-252b ≥98% (by HPLC)
  ENZOBMLEI2340100
 :  

 

  • Pk:
    100 µG
  • Seq:
    1286

 

 

 :
 :453.5 g/mol
 :Refrigerator
 :99570-78-2
 :C₂₆H₁₉N₃O₅