Order Entry
Czech Republic
ContactUsLinkComponent
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
Catalog # ENZOBMLDS0560050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
Catalog # ENZOBMLDS0560050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

About this item

 :
 :336.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
CAS Number:82948-88-7
Formula:C₂₀H₃₂O₄